Tisk…

4-Methoxy-N-methylbenzylamine 98%

BLDPBD29448-1GEA 371 CZK
BLDPBD29448-1G BLDPBD29448-100G BLDPBD29448-10G BLDPBD29448-25G BLDPBD29448-5G
4-Methoxy-N-methylbenzylamine 98%
4-Methoxy-N-methylbenzylamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₁₃NO
MW: 151,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115407
CAS číslo: 702-24-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR