Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate 95%
|
|
BLDPBD29460-25GEA
14430
CZK
BLDPBD29460-25G
BLDPBD29460-5G
BLDPBD29460-1G
BLDPBD29460-0.025G
BLDPBD29460-250MG
Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₁H₁₂N₂O₂ MW: 204,2300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02187526 CAS číslo: 70705-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...