Tisk…

4-Isoquinolinecarboxylic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD29478-25GEA 23350 CZK
BLDPBD29478-25G BLDPBD29478-250MG BLDPBD29478-5G BLDPBD29478-10G BLDPBD29478-1G
4-Isoquinolinecarboxylic acid 98%
4-Isoquinolinecarboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₇NO₂
MW: 173,1681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094344
CAS číslo: 7159-36-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR