Tisk…

N-Cyano-N-phenyl-p-toluenesulphonamide 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD294888-250MGEA 741 CZK
BLDPBD294888-250MG BLDPBD294888-1G BLDPBD294888-100MG BLDPBD294888-5G
N-Cyano-N-phenyl-p-toluenesulphonamide 95%
N-Cyano-N-phenyl-p-toluenesulphonamide

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₄H₁₂N₂O₂S
MW: 272,322 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00159358
CAS číslo: 55305-43-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR