2-Acetyl-3-(Boc-amino)pyrazine 98%
|
|
BLDPBD296989-1GEA
4340
CZK
BLDPBD296989-1G
BLDPBD296989-100MG
BLDPBD296989-250MG
2-Acetyl-3-(Boc-amino)pyrazine 98%
2-Acetyl-3-(Boc-amino)pyrazine
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₁H₁₅N₃O₃ MW: 237,255 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28963469 CAS číslo: 1799412-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...