Tisk…

2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD301714-1GEA 3800 CZK
BLDPBD301714-1G BLDPBD301714-100MG BLDPBD301714-5G BLDPBD301714-250MG
2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one 98%
2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₆H₃ClN₂OS
MW: 186,619 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19382468
CAS číslo: 1245811-20-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR