Tisk…

Methyl 3-chloro-5-hydroxypyridine-2-carboxylate 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD302971-100MGEA 4850 CZK
BLDPBD302971-100MG BLDPBD302971-250MG
Methyl 3-chloro-5-hydroxypyridine-2-carboxylate 95%
Methyl 3-chloro-5-hydroxypyridine-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₆ClNO₃
MW: 187,58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257707
CAS číslo: 1256811-09-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR