Tisk…

4-Amino-1-methylpiperidine 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD3037-1GEA 171 CZK
BLDPBD3037-1G BLDPBD3037-25G BLDPBD3037-10G BLDPBD3037-5G
4-Amino-1-methylpiperidine 97%
4-Amino-1-methylpiperidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₁₄N₂
MW: 114,1888 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01570549
CAS číslo: 41838-46-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR