Tisk…

4'-Methyl-3'-nitroacetanilide 95+%

BLDPBD305729-1GEA 314 CZK
BLDPBD305729-1G BLDPBD305729-25G BLDPBD305729-5G
4'-Methyl-3'-nitroacetanilide 95+%
4'-Methyl-3'-nitroacetanilide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₁₀N₂O₃
MW: 194,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024277
CAS číslo: 2719-14-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR