Trypan blue 0.7 (dye content)
|
|
BLDPBD306386-25GEA
1200
CZK
BLDPBD306386-25G
BLDPBD306386-5G
Trypan blue 0.7 (dye content)
Trypan blue
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₃₄H₂₄N₆Na₄O₁₄S₄ MW: 960,82 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003969 CAS číslo: 72-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...