2-Bromo-4,5,6-trichloroaniline 98%
|
|
BLDPBD3116-25GEA
3200
CZK
BLDPBD3116-25G
BLDPBD3116-5G
BLDPBD3116-10G
BLDPBD3116-1G
2-Bromo-4,5,6-trichloroaniline 98%
2-Bromo-4,5,6-trichloroaniline
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₆H₃BrF₃N MW: 225,9900 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00013388 CAS číslo: 122375-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...