Tisk…

Ethyl (2-bromophenyl)acetate 98%

BLDPBD315464-25GEA 2770 CZK
BLDPBD315464-25G BLDPBD315464-10G BLDPBD315464-1G BLDPBD315464-5G BLDPBD315464-100G
Ethyl (2-bromophenyl)acetate 98%
Ethyl (2-bromophenyl)acetate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₁BrO₂
MW: 243,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051921
CAS číslo: 2178-24-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR