6-Hydroxy-2,2-dimethyl-2,3-dihydro-4H-chromen-4-one 97%
|
|
BLDPBD315540-5GEA
8950
CZK
BLDPBD315540-5G
BLDPBD315540-250MG
BLDPBD315540-100MG
BLDPBD315540-1G
6-Hydroxy-2,2-dimethyl-2,3-dihydro-4H-chromen-4-one 97%
6-Hydroxy-2,2-dimethyl-2,3-dihydro-4H-chromen-4-one
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₁H₁₂O₃ MW: 192,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03830209 CAS číslo: 31366-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...