2-(3-Nitrophenyl)-1,3,4-oxadiazole 98%
|
|
BLDPBD315716-250MGEA
485
CZK
BLDPBD315716-250MG
BLDPBD315716-1G
BLDPBD315716-5G
2-(3-Nitrophenyl)-1,3,4-oxadiazole 98%
2-(3-Nitrophenyl)-1,3,4-oxadiazole
Storage condition: Sealed in dry, 2-8°C
Vzorec:
C₈H₅N₃O₃ MW: 191,144 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00085145 CAS číslo: 5565-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...