Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%
|
|
BLDPBD315793-250MGEA
1860
CZK
BLDPBD315793-250MG
BLDPBD315793-5G
BLDPBD315793-100MG
BLDPBD315793-1G
Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₀H₉ClN₂O₂ MW: 224,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09864797 CAS číslo: 67625-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...