Tisk…

2,2'-Dibenzothiazole disulphide 96%

BLDPBD31632-25GEA 171 CZK
BLDPBD31632-25G
2,2'-Dibenzothiazole disulphide 96%
2,2'-Dibenzothiazole disulphide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₄H₈N₂S₄
MW: 332,4867 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022874
CAS číslo: 120-78-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR