Tisk…

Ethyl-6-bromo-4-hydroxyquinoline-3-carboxylate 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD31658-1GEA 200 CZK
BLDPBD31658-1G BLDPBD31658-25G BLDPBD31658-10G BLDPBD31658-250MG BLDPBD31658-100G BLDPBD31658-5G
Ethyl-6-bromo-4-hydroxyquinoline-3-carboxylate 95%
Ethyl-6-bromo-4-hydroxyquinoline-3-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₂H₁₀BrNO₃
MW: 296,1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173362
CAS číslo: 122794-99-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR