Tisk…

1H-Imidazol-1-amine hydrochloride 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD317960-100MGEA 2090 CZK
BLDPBD317960-100MG BLDPBD317960-5G BLDPBD317960-1G BLDPBD317960-250MG
1H-Imidazol-1-amine hydrochloride 97%
1H-Imidazol-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₃H₅N₃·ClH
MW: 119,553 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26390998
CAS číslo: 83279-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR