5-Chloro-2-hydroxybenzonitrile 97%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD31820-25GEA
5130
CZK
BLDPBD31820-25G
BLDPBD31820-5G
BLDPBD31820-10G
BLDPBD31820-250MG
BLDPBD31820-1G
5-Chloro-2-hydroxybenzonitrile 97%
5-Chloro-2-hydroxybenzonitrile
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₇H₄ClNO MW: 153,5700 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00219003 CAS číslo: 13589-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...