Tisk…

4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD32078-5GEA 542 CZK
BLDPBD32078-5G BLDPBD32078-10G BLDPBD32078-100G BLDPBD32078-1G BLDPBD32078-25G
4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%
4-Hydroxy-6-(trifluoromethyl)pyrimidine

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₅H₃F₃N₂O
MW: 164,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192530
CAS číslo: 1546-78-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR