Tisk…

4-Propoxyphenylboronic acid 97%

BLDPBD32389-5GEA 1140 CZK
BLDPBD32389-5G BLDPBD32389-25G BLDPBD32389-1G BLDPBD32389-10G
4-Propoxyphenylboronic acid 97%
4-Propoxyphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₁₃BO₃
MW: 180,01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03427189
CAS číslo: 186497-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR