Tisk…

Isoquinoline-7-amine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD32792-250MGEA 684 CZK
BLDPBD32792-250MG BLDPBD32792-100MG BLDPBD32792-5G BLDPBD32792-1G
Isoquinoline-7-amine 98%
Isoquinoline-7-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₉H₈N₂
MW: 144,1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669932
CAS číslo: 23707-37-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR