Tisk…

Imidazo[1,2-a]pyridine 95%

BLDPBD32995-25GEA 3910 CZK
BLDPBD32995-25G BLDPBD32995-100G BLDPBD32995-1G BLDPBD32995-5G BLDPBD32995-10G
Imidazo[1,2-a]pyridine 95%
Imidazo[1,2-a]pyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₇H₆N₂
MW: 118,1359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005553
CAS číslo: 274-76-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR