Tisk…

4-Chloro-3-nitrophenylboronic acid 98%

BLDPBD3315-250MGEA 285 CZK
BLDPBD3315-250MG BLDPBD3315-1G
4-Chloro-3-nitrophenylboronic acid 98%
4-Chloro-3-nitrophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₅BClNO₄
MW: 201,37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02258950
CAS číslo: 151169-67-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR