Tisk…

2-Methoxy-6-quinolineboronic acid pinacol ester 98%

BLDPBD331880-250MGEA 1890 CZK
BLDPBD331880-250MG BLDPBD331880-1G BLDPBD331880-100MG
2-Methoxy-6-quinolineboronic acid pinacol ester 98%
2-Methoxy-6-quinolineboronic acid pinacol ester

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₆H₂₀BNO₃
MW: 285,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383956
CAS číslo: 1201644-36-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR