Methyl-4-bromo-3-hydroxythiophene-2-carboxylate 95%
|
|
BLDPBD3347-5GEA
7670
CZK
BLDPBD3347-5G
BLDPBD3347-250MG
BLDPBD3347-100MG
BLDPBD3347-25G
BLDPBD3347-1G
Methyl-4-bromo-3-hydroxythiophene-2-carboxylate 95%
Methyl-4-bromo-3-hydroxythiophene-2-carboxylate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₆H₅BrO₃S MW: 237,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052081 CAS číslo: 95201-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...