Tisk…

2-Bromo-5-(trifluoromethyl)phenol 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD33579-1GEA 399 CZK
BLDPBD33579-1G BLDPBD33579-100G BLDPBD33579-0.025G BLDPBD33579-5G BLDPBD33579-10G BLDPBD33579-25G
2-Bromo-5-(trifluoromethyl)phenol 95%
2-Bromo-5-(trifluoromethyl)phenol

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₄BrF₃O
MW: 241,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261190
CAS číslo: 402-05-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR