Tisk…

2-Aminooxazole 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD33700-1GEA 513 CZK
BLDPBD33700-1G BLDPBD33700-10G BLDPBD33700-5G BLDPBD33700-250MG BLDPBD33700-25G
2-Aminooxazole 97%
2-Aminooxazole
Vzorec: C₃H₄N₂O
MW: 84,10 g/mol
MDL Number: MFCD07364485
CAS číslo: 4570-45-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR