Tisk…

(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene 95%

BLDPBD337791-100MGEA 969 CZK
BLDPBD337791-100MG BLDPBD337791-5G BLDPBD337791-250MG BLDPBD337791-1G
(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene 95%
(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₂₂H₁₆Br₂O₂
MW: 472,169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06654379
CAS číslo: 117745-41-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR