2-Chloro-4-fluorophenylhydrazine hydrochloride 98%
|
|
BLDPBD33807-10GEA
1200
CZK
BLDPBD33807-10G
BLDPBD33807-1G
BLDPBD33807-5G
BLDPBD33807-25G
BLDPBD33807-100G
2-Chloro-4-fluorophenylhydrazine hydrochloride 98%
2-Chloro-4-fluorophenylhydrazine hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₆H₇Cl₂FN₂ MW: 197,0376 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03788524 CAS číslo: 497959-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...