Tisk…

2-Bromo-7-methoxynaphthalene 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD338145-1GEA 6500 CZK
BLDPBD338145-1G BLDPBD338145-250MG BLDPBD338145-100MG
2-Bromo-7-methoxynaphthalene 95%
2-Bromo-7-methoxynaphthalene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₉BrO
MW: 237,093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17012538
CAS číslo: 200875-36-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR