Tisk…

8-Methyl-3,4-dihydroquinolin-2(1H)-one 95%

BLDPBD338146-100MGEA 3340 CZK
BLDPBD338146-100MG BLDPBD338146-250MG
8-Methyl-3,4-dihydroquinolin-2(1H)-one 95%
8-Methyl-3,4-dihydroquinolin-2(1H)-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464327
CAS číslo: 20151-47-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR