Tisk…

2,4,6-Trichloropyrimidine-5-carbaldehyde 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD33832-1GEA 200 CZK
BLDPBD33832-1G BLDPBD33832-0.025G BLDPBD33832-5G BLDPBD33832-25G
2,4,6-Trichloropyrimidine-5-carbaldehyde 97%
2,4,6-Trichloropyrimidine-5-carbaldehyde

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₅HCl₃N₂O
MW: 211,43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02257700
CAS číslo: 50270-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR