Tisk…

Ethyl-2-aminobenzo[d]thiazole-6-carboxylate 98%

BLDPBD33863-5GEA 684 CZK
BLDPBD33863-5G BLDPBD33863-100G BLDPBD33863-25G BLDPBD33863-500G BLDPBD33863-1G BLDPBD33863-10G
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate 98%
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₁₀N₂O₂S
MW: 222,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00102724
CAS číslo: 50850-93-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR