Tisk…

2-Formyl-4-picoline 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD33996-100GEA 61760 CZK
BLDPBD33996-100G BLDPBD33996-25G BLDPBD33996-5G BLDPBD33996-250MG BLDPBD33996-1G
2-Formyl-4-picoline 98%
2-Formyl-4-picoline

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₇NO
MW: 121,1366 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07367933
CAS číslo: 53547-60-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR