2-Iodo-4-(trifluoromethyl)benzoic acid 98%
|
|
BLDPBD34041-250MGEA
456
CZK
BLDPBD34041-250MG
BLDPBD34041-1G
BLDPBD34041-5G
2-Iodo-4-(trifluoromethyl)benzoic acid 98%
2-Iodo-4-(trifluoromethyl)benzoic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₈H₄F₃IO₂ MW: 316,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06408817 CAS číslo: 54507-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...