Tisk…

Imidazo[1,2-b]pyridazine 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD3410-25GEA 855 CZK
BLDPBD3410-25G BLDPBD3410-0.025G BLDPBD3410-1G BLDPBD3410-100G BLDPBD3410-5G BLDPBD3410-10G
Imidazo[1,2-b]pyridazine 95%
Imidazo[1,2-b]pyridazine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₅N₃
MW: 119,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782103
CAS číslo: 766-55-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR