3-Amino-3-(3-methylphenyl)propionic acid 98%
|
|
BLDPBD34611-1GEA
627
CZK
BLDPBD34611-1G
BLDPBD34611-5G
BLDPBD34611-250MG
3-Amino-3-(3-methylphenyl)propionic acid 98%
3-Amino-3-(3-methylphenyl)propionic acid
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₀H₁₃NO₂ MW: 179,2157 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01871312 CAS číslo: 68208-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...