Tisk…

3-Amino-3-(2-chlorophenyl)propionic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD34612-1GEA 969 CZK
BLDPBD34612-1G BLDPBD34612-5G BLDPBD34612-250MG
3-Amino-3-(2-chlorophenyl)propionic acid 98%
3-Amino-3-(2-chlorophenyl)propionic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01871299
CAS číslo: 68208-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR