Tisk…

4-Cyano-3-fluorophenylboronic acid 97%

BLDPBD35066-5GEA 1520 CZK
BLDPBD35066-5G BLDPBD35066-10G BLDPBD35066-250MG BLDPBD35066-1G BLDPBD35066-100G BLDPBD35066-0.025G BLDPBD35066-25G
4-Cyano-3-fluorophenylboronic acid 97%
4-Cyano-3-fluorophenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₇H₅BFNO₂
MW: 164,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411549
CAS číslo: 843663-18-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR