Tisk…

5-Bromo-2,3-difluoropyridine 98%

BLDPBD35345-1GEA 285 CZK
BLDPBD35345-1G BLDPBD35345-10G BLDPBD35345-25G BLDPBD35345-100G BLDPBD35345-5G
5-Bromo-2,3-difluoropyridine 98%
5-Bromo-2,3-difluoropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₅H₂BrF₂N
MW: 193,98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659525
CAS číslo: 89402-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR