Tisk…

Methyl glycolate 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD35524-1000GEA 2510 CZK
BLDPBD35524-1000G BLDPBD35524-100G BLDPBD35524-500G BLDPBD35524-25G BLDPBD35524-10G BLDPBD35524-5G
Methyl glycolate 98%
Methyl glycolate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₃H₆O₃
MW: 90,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004667
CAS číslo: 96-35-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR