Tisk…

5-Methyl-3-pyridineboronic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD3668-250MGEA 228 CZK
BLDPBD3668-250MG BLDPBD3668-1G
5-Methyl-3-pyridineboronic acid 98%
5-Methyl-3-pyridineboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₆H₈BNO₂
MW: 136,95 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03428528
CAS číslo: 173999-18-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR