Tisk…

4-Amino-2-methoxypyridine 98%

BLDPBD3694-5GEA 314 CZK
BLDPBD3694-5G BLDPBD3694-0.5G BLDPBD3694-1G BLDPBD3694-25G BLDPBD3694-100G BLDPBD3694-10G BLDPBD3694-500G
4-Amino-2-methoxypyridine 98%
4-Amino-2-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₈N₂O
MW: 124,1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738657
CAS číslo: 20265-39-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR