Methyl-2-aminoisonicotinate 98%
|
|
BLDPBD36970-25GEA
1460
CZK
BLDPBD36970-25G
BLDPBD36970-100G
BLDPBD36970-10G
BLDPBD36970-1G
BLDPBD36970-5G
Methyl-2-aminoisonicotinate 98%
Methyl-2-aminoisonicotinate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₇H₈N₂O₂ MW: 152,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039316 CAS číslo: 6937-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...