α,3-Dibromo-4-fluorotoluene 98%
|
|
BLDPBD37539-10GEA
1290
CZK
BLDPBD37539-10G
BLDPBD37539-5G
BLDPBD37539-25G
α,3-Dibromo-4-fluorotoluene 98%
α,3-Dibromo-4-fluorotoluene
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₇H₅Br₂F MW: 267,92 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09037367 CAS číslo: 78239-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...