4-(Aminomethyl)phenylboronic acid hydrochloride 97%
|
|
BLDPBD3767-1GEA
1060
CZK
BLDPBD3767-1G
BLDPBD3767-250MG
BLDPBD3767-10G
BLDPBD3767-5G
BLDPBD3767-25G
BLDPBD3767-100MG
4-(Aminomethyl)phenylboronic acid hydrochloride 97%
4-(Aminomethyl)phenylboronic acid hydrochloride
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₇H₁₁BClNO₂ MW: 187,43 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01632199 CAS číslo: 75705-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...