3-(Aminomethyl)phenylboronic acid hydrochloride 95%
|
|
BLDPBD3768-1GEA
1940
CZK
BLDPBD3768-1G
BLDPBD3768-250MG
BLDPBD3768-5G
BLDPBD3768-25G
3-(Aminomethyl)phenylboronic acid hydrochloride 95%
3-(Aminomethyl)phenylboronic acid hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₇H₁₁BClNO₂ MW: 187,43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02093050 CAS číslo: 352525-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...