7-Hydroxy-2-methylchromone 95%
BLDPBD396666-100MGEA
1520
CZK
BLDPBD396666-100MG
BLDPBD396666-250MG
7-Hydroxy-2-methylchromone 95%
7-Hydroxy-2-methylchromone
Vzorec:
C₁₀H₈O₃ MW: 176,1700 g/mol |
MDL Number:
MFCD00208225 CAS číslo: 6320-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...