Tisk…

2,4,5-Trichlorobenzoic acid 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD41741-1GEA 513 CZK
BLDPBD41741-1G BLDPBD41741-5G BLDPBD41741-10G BLDPBD41741-0.025G BLDPBD41741-100G BLDPBD41741-250MG BLDPBD41741-25G BLDPBD41741-100MG
2,4,5-Trichlorobenzoic acid 95%
2,4,5-Trichlorobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₃Cl₃O₂
MW: 225,4600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01662044
CAS číslo: 50-82-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR